XSelect Peptide CSH C18 OBD Prep Column, 130Å, 5 µm, 50 mm X 100 mm, 1K - 15K 1 * 1 items
Catalog # WATE186007027
Supplier: WATERS
XSelect Peptide CSH C18 OBD Prep Column, 130Å, 5 µm, 50 mm X 100 mm, 1K - 15K 1 * 1 items
Catalog # WATE186007027
Supplier: WATERS
Frequently Bought Together
Avantor® ACE® C8, HPLC Columns, 5 µm
Supplier: Avantor
VWR®, Storage Boxes for Sample Vials and Bottles, PP
Supplier: VWR Collection
VWR®, Screw Neck Vials ND15
Supplier: VWR Collection
Avantor® ACE® C18, HPLC Columns, 3 µm
Supplier: Avantor
Avantor® Alltima, HPLC Columns, C18
Supplier: Avantor
Avantor® ACE® CN, HPLC Columns, 5 µm
Supplier: Avantor
VWR® Hitachi LaChromUltra II, LaChrom HPLC Columns
Supplier: VWR Collection
Avantor® ACE® C18, HPLC Columns, 5 µm
Supplier: Avantor
Avantor® ACE®, Analytical / Semi-Preparative Column Coupler
Supplier: Avantor
VWR®, Crimp Neck Vials ND11
Supplier: VWR Collection
Valves for syringes with valves
Supplier: Trajan Scientific and Medical
Snap ring vials, N 11
Supplier: MACHEREY-NAGEL
Glass snap top vials, 2 ml, level 2 high-throughput applications, SureSTART™
Supplier: Thermo Fisher Scientific
Accessories for SCAT Safety Caps
Supplier: S.C.A.T.