XSelect CSH Fluoro-Phenyl Column, 130Å, 5 µm, 4.6 mm X 75 mm 1 * 1 items
Catalog # WATE186005343
Supplier: WATERS
XSelect CSH Fluoro-Phenyl Column, 130Å, 5 µm, 4.6 mm X 75 mm 1 * 1 items
Catalog # WATE186005343
Supplier: WATERS
Frequently Bought Together
Avantor® Alltima, HPLC Columns, C18
Supplier: Avantor
Avantor® Apollo C18, HPLC Columns
Supplier: Avantor
PK20 PRECLEANED ASSEMBLED 1/4IN BRASS 1 * 1 items
Supplier: Merck
XELA UHPLC Columns, Biphenyl
Supplier: ISERA
UPC2 STANDARDS KIT 1 * 1 KIT
Supplier: WATERS
QuEChERS mixes, clean-up, CHROMABOND®
Supplier: MACHEREY-NAGEL
SureSTART™ Glass Inserts for 2 ml Vials, Level 3 High Performance Applications
Supplier: Thermo Fisher Scientific
HPLC columns, Waters Spherisorb®
Supplier: WATERS
Cuvettes for fluorescence<br />measurements
Supplier: VARIAN
Screw neck vials, N 9
Supplier: MACHEREY-NAGEL
Microvolume syringes 10 µl
Supplier: Trajan Scientific and Medical
XELA UHPLC Columns, HILIC-Diol
Supplier: ISERA
Calibration standard, single element, silicon for AAS and ICP-OES, pure grade
Supplier: PerkinElmer
Ion exclusion chromatography columns, KC-811
Supplier: Resonac