XBridge BEH HILIC OBD Prep Column, 130Å, 5 µm, 50 mm X 50 mm 1 * 1 items
Catalog # WATE186004732
Supplier: WATERS
XBridge BEH HILIC OBD Prep Column, 130Å, 5 µm, 50 mm X 50 mm 1 * 1 items
Catalog # WATE186004732
Supplier: WATERS
Frequently Bought Together
Avantor® Allsep Anion, Anion Exchange HPLC Columns
Supplier: Avantor
AccQ-Tag Hydrolysates Chemistry Kit - Automation 1 * 1 KIT
Supplier: WATERS
PROPAC 3R SAX, 3UM, 2.0X50MM 1 * 1 items
Supplier: Thermo Fisher Scientific
TSKgel® gel filtration columns, PW-type
Supplier: TOSOH Bioscience
Chromatography Tubing, Stainless Steel
Supplier: Avantor
Digestion tubes, DigiTUBE®
Supplier: PerkinElmer
Flash chromatography cartridges, CHROMABOND® Flash RS
Supplier: MACHEREY-NAGEL
Chromatogram immersion device for TLC plates
Supplier: CAMAG
OUTLET MIST FILTER NW25 1 * 1 items
Supplier: Agilent
CP-SIL 5CB-MS FS 30X.32(1.0) 1 * 1 items
Supplier: VARIAN
Coated polysaccharide columns, normal phase, CHIRALPAK®
Supplier: CHIRAL TECHNOLOGIES
HPLC Separation Column EC 50/4.6 NUCLEOSIL 100-5 NO2 Length: 50 mm, ID: 4.6 mm 1 * 1 items
Supplier: MACHEREY-NAGEL