Atlantis T3 OBD Prep Column, 100Å, 10 µm, 50 mm X 100 mm 1 * 1 items
Catalog # WATE186004084
Supplier: WATERS
Atlantis T3 OBD Prep Column, 100Å, 10 µm, 50 mm X 100 mm 1 * 1 items
Catalog # WATE186004084
Supplier: WATERS
Frequently Bought Together
Avantor® Alltima, HPLC Columns, C18
Supplier: Avantor
Avantor® ACE® C18, HPLC Columns, 3 µm
Supplier: Avantor
VWR®, Screw Neck Headspace Vials ND18
Supplier: VWR Collection
Avantor® ACE® C18, HPLC Columns, 5 µm
Supplier: Avantor
VWR®, Short Thread Vials ND9
Supplier: VWR Collection
VWR®, Inserts for Screw Neck Vials ND8
Supplier: VWR Collection
EPA screw neck vials, N 24
Supplier: MACHEREY-NAGEL
Florisil® 60-100 mesh for chromatography
Supplier: VWR Chemicals
VWR®, Crimp Neck Vials ND11
Supplier: VWR Collection
Air valves for safetycaps generation 2.0
Supplier: S.C.A.T.
VWR®, Inserts for Short Thread Vials ND9
Supplier: VWR Collection
Screw caps for glass screw top vials, 9 mm, level 1 everyday analysis, SureSTART™
Supplier: Thermo Fisher Scientific
Inlet liners
Supplier: Agilent
Screw caps for certified screw top vials
Supplier: Agilent