Order Entry
Ireland
ContactUsLinkComponent
ACQUITY UPLC BEH HILIC Column, 130Å, 1.7 µm, 1 mm X 150 mm 1 * 1 items
Catalog # WATE186003459
Supplier:  WATERS
ACQUITY UPLC BEH HILIC Column, 130Å, 1.7 µm, 1 mm X 150 mm 1 * 1 items
Catalog # WATE186003459
Supplier:  WATERS
Frequently Bought Together