XBridge BEH Shield RP18 Sentry Guard Cartridge, 130Å, 3.5 µm, 3 mm X 20 mm 1 * 2 items
Catalog # WATE186003069
Supplier: WATERS
XBridge BEH Shield RP18 Sentry Guard Cartridge, 130Å, 3.5 µm, 3 mm X 20 mm 1 * 2 items
Catalog # WATE186003069
Supplier: WATERS
Frequently Bought Together
VWR® HPLC Microsyringes, Rheodyne
Supplier: VWR Collection
VWR®, Screw Caps for Short Thread Vials ND9
Supplier: VWR Collection
TLC plates, silica gel 60G, glass backed
Supplier: Merck
Exhaust filters, V3.0
Supplier: S.C.A.T.
Reducing tubing connectors, b.safe
Supplier: Bohlender
VWR®, Short Thread Vials ND9
Supplier: VWR Collection
VWR®, Crimp Neck Headspace Vials ND20
Supplier: VWR Collection
Accessories for connection system GLS 80
Supplier: DWK Life Sciences
Silica gel, Pore size: 60 Å 0.063-0.200 mm (70-230 mesh ASTM) for column chromatography, Millipore®
Supplier: MERCK PRODUCTION CHEMICALS
VWR® ACE® Excel® C18 HPLC/UHPLC Columns, Analytical, 2 µm
Supplier: Avantor
CAP SCREW 24MM WHITE PP 24-400 PE FOAM / WHITE PTFE CLOSE TOP 100/PACK 1 * 100 items
Supplier: Thermo Fisher Scientific
SureSTART™ Glass Inserts for 2 ml Vials, Level 3 High Performance Applications
Supplier: Thermo Fisher Scientific
Precision screw caps, 18 mm, level 3 high performance applications, SureSTART™
Supplier: Thermo Fisher Scientific
Certified screw top vials and closures
Supplier: Agilent
Screw caps for certified screw top vials
Supplier: Agilent