XBridge BEH C18 Sentry Guard Cartridge, 130Å, 5 µm, 2.1 mm X 10 mm 1 * 2 items
Catalog # WATE186003062
Supplier: WATERS
XBridge BEH C18 Sentry Guard Cartridge, 130Å, 5 µm, 2.1 mm X 10 mm 1 * 2 items
Catalog # WATE186003062
Supplier: WATERS
Frequently Bought Together
Avantor® Allsep Anion, Anion Exchange HPLC Columns
Supplier: Avantor
Avantor® ACE® C18, HPLC Columns, 5 µm
Supplier: Avantor
Screw neck vials, N 13
Supplier: MACHEREY-NAGEL
Avantor® Alltima, HPLC Columns, C18
Supplier: Avantor
SYRINGE W/O NEEDLE 75 RN 5UL CALIBRATED 1 * 1 items
Supplier: HAMILTON BONADUZ
Accessories for 8453 uv-vis cuvettes
Supplier: Agilent
100R-C/T-GT-0.47C SYRINGE CALIBRATED 1 * 1 items
Supplier: Trajan Scientific and Medical
Closures for headspace vials
Supplier: PerkinElmer
XELA UHPLC Columns, C18-AQ-PolE
Supplier: ISERA
GC septa
Supplier: Trajan Scientific and Medical
SYRINGE 10CC PLAIN 1 * 1 items
Supplier: WATERS
Connectors, tubing, miniature couplings
Supplier: Bohlender
Calibration standard, single element, potassium for AAS and ICP-OES, pure grade
Supplier: PerkinElmer
LC fittings, ferrules and unions
Supplier: Agilent