XBridge Premier Protein BEH C4 300 Å 2.5 µm, 4.6 x 50 mm with MassPREP Protein Mix Standard 1 * 1 items
Catalog # WATE176005113
Supplier: WATERS
XBridge Premier Protein BEH C4 300 Å 2.5 µm, 4.6 x 50 mm with MassPREP Protein Mix Standard 1 * 1 items
Catalog # WATE176005113
Supplier: WATERS
Frequently Bought Together
VWR®, Screw Caps for Short Thread Vials ND9
Supplier: VWR Collection
VWR®, Short Thread Vials ND9
Supplier: VWR Collection
VWR®, Crimp Caps for Crimp Neck Vials ND11
Supplier: VWR Collection
SafetyCaps
Supplier: S.C.A.T.
Screw caps for screw neck vials, N 13
Supplier: MACHEREY-NAGEL
Glass capillaries and graduated micropipettes, disposable
Supplier: CAMAG
VWR®, Snap Cap Vials
Supplier: VWR Collection
Headspace crimp caps with 20 mm septa
Supplier: Agilent
Air valves for safetycaps generation 2.0
Supplier: S.C.A.T.
Glass crimp top vials, 2 ml, level 2 high-throughput applications, SureSTART™
Supplier: Thermo Fisher Scientific
TLC plates, silica gel 60, unmodified layers, plastic sheets
Supplier: Merck
Certified screw top vials and closures
Supplier: Agilent
Gold standard syringes for autosampler
Supplier: Agilent
PTFE/rubber laminated discs
Supplier: Thermo Fisher Scientific