Order Entry
Ireland
ContactUsLinkComponent
CORTECS C18 Column, 90Å. 2.7 µm, 2.1 x 50 mm, with MarvelX SS UHPLC connection system, 125 µm x 400 mm Kit 1 * 2 items
Catalog # WATE176004955
Supplier:  WATERS
CORTECS C18 Column, 90Å. 2.7 µm, 2.1 x 50 mm, with MarvelX SS UHPLC connection system, 125 µm x 400 mm Kit 1 * 2 items
Catalog # WATE176004955
Supplier:  WATERS
Frequently Bought Together