ACQUITY UPLC CSH Phenyl-Hexyl Column, 130Å, 1.7 µm, 2.1 mm X 30 mm 1 * 3 items
Catalog # WATE176002162
Supplier: WATERS
ACQUITY UPLC CSH Phenyl-Hexyl Column, 130Å, 1.7 µm, 2.1 mm X 30 mm 1 * 3 items
Catalog # WATE176002162
Supplier: WATERS
Frequently Bought Together
Avantor® ACE® C8, HPLC Columns, 5 µm
Supplier: Avantor
Avantor® ACE® C18, HPLC Columns, 5 µm
Supplier: Avantor
Capillaries for LC systems
Supplier: Agilent
MOISTURE/HYDROCARBON TRAP CLICK-ON 1 * 1 items
Supplier: PerkinElmer
J.T.Baker® BAKERBOND spe™, SPE Glass Columns with PTFE Frits
Supplier: Avantor
FlashPure Select Silica Cartridges
Supplier: BUCHI
SymLine Automatic Switching for Container
Supplier: S.C.A.T.
20Mm Borate Buffer Ph9.3 100Ml 1 * 100 mL
Supplier: Agilent
REPROSPHER 100 CN-DE, 5 µM 100 X 4.6 MM 1 * 1 items
Supplier: DR MAISCH
FlashPure EcoFlex Silica Cartridges
Supplier: BUCHI
PTFE/rubber laminated discs
Supplier: Thermo Fisher Scientific
500UL SAMPLE LOOP 1 * 1 items
Supplier: PerkinElmer