Order Entry
Ireland
ContactUsLinkComponent
SunFire C18 OBD Prep Column, 100Å, 10 µm, 30 mm X 250 mm 1 * 1 items
Catalog # WATE186002671
Supplier:  WATERS
SunFire C18 OBD Prep Column, 100Å, 10 µm, 30 mm X 250 mm 1 * 1 items
Catalog # WATE186002671
Supplier:  WATERS
Frequently Bought Together