SunFire C18 OBD Prep Column, 100Å, 10 µm, 30 mm X 150 mm 1 * 1 items
WATE186002670
: WATERS
Frequently Bought Together
Accessories for electronic crimping tools
: MACHEREY-NAGEL
Screw neck vials
: WATERS
Round canisters
: S.C.A.T.
Macro cuvettes
: Agilent
SCV gas sampling bags, Tedlar®
: Merck
SureSTART™ Crimp Caps, 11 mm, Level 2 High-Throughput Applications
: Thermo Fisher Scientific
Accessories for waste tubes, b.safe
: Bohlender
Accessories for glass block manifolds, HyperSep™
: Thermo Fisher Scientific
Manual GC syringes
: Thermo Fisher Scientific