XTerra Shield RP8 OBD Prep Column, 125Å, 10 µm, 19 mm X 150 mm 1 * 1 items
WATE186002258
: WATERS
Frequently Bought Together
VWR®, Crimp Neck Vials ND11
: VWR Collection
VWR®, Crimp Neck Headspace Vials ND20
: VWR Collection
Florisil® 60-100 mesh for chromatography
: VWR Chemicals
VWR®, Short Thread Vials ND9
: VWR Collection
VWR®, Screw Neck Vials ND8
: VWR Collection
VWR®, Screw Caps for Screw Neck Vials ND24
: VWR Collection
VWR®, Screw Neck Vials ND13
: VWR Collection
VWR®, Screw Caps for Short Thread Vials ND9
: VWR Collection
VWR®, Screw Neck Vials ND24
: VWR Collection
VWR®, Snap Cap Vials
: VWR Collection
Exhaust filters, V3.0
: S.C.A.T.
VWR®, Screw Neck Vials ND10
: VWR Collection
VWR® Standard Micro Syringes
: VWR Collection
Air valves for safetycaps generation 2.0
: S.C.A.T.
VWR®, Inserts for Short Thread Vials ND9
: VWR Collection
Crimp top vials and closures, 11 mm
: Agilent