Order Entry
Ireland
ContactUsLinkComponent
LCGC Certified Amber Glass 15 x 45 mm Screw Neck Vial, with Cap and PTFE/Silicone Septum, 4 mL Volume, 1 * 100 items
Catalog # WATE186001133C
Supplier:  WATERS
LCGC Certified Amber Glass 15 x 45 mm Screw Neck Vial, with Cap and PTFE/Silicone Septum, 4 mL Volume, 1 * 100 items
Catalog # WATE186001133C
Supplier:  WATERS
Frequently Bought Together