Deactivated Clear Glass 15 x 45 mm Screw Neck Vial, with Cap and PTFE/Silicone Septum, 4 mL Volume, 1 * 100 items
Catalog # WATE186000838DV
Supplier: WATERS
Deactivated Clear Glass 15 x 45 mm Screw Neck Vial, with Cap and PTFE/Silicone Septum, 4 mL Volume, 1 * 100 items
Catalog # WATE186000838DV
Supplier: WATERS
Frequently Bought Together
VWR®, Crimp Neck Vials ND11
Supplier: VWR Collection
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR®, Short Thread Vials ND9
Supplier: VWR Collection
VWR®, Screw Neck Headspace Vials ND18
Supplier: VWR Collection
VWR®, Manual Crimper and Decapper
Supplier: VWR Collection
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Headspace crimp caps with 20 mm septa
Supplier: Agilent
Exhaust filters, V3.0
Supplier: S.C.A.T.
VWR®, Crimp Caps for Crimp Neck Vials ND11
Supplier: VWR Collection
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
VWR®, Screw Neck Vials ND13
Supplier: VWR Collection
Laboratory bottles, narrow neck, with screw cap, DURAN®
Supplier: DWK Life Sciences
Ethanol, GPR RECTAPUR®
Supplier: VWR Chemicals