Solid Cap for 22 mL Vial, PTFE/Silicone Liner, 1 * 100 items
Catalog # WATE186004109
Supplier: WATERS
Solid Cap for 22 mL Vial, PTFE/Silicone Liner, 1 * 100 items
Catalog # WATE186004109
Supplier: WATERS
Frequently Bought Together
Avantor® ACE® C18-PFP, HPLC Columns, 5 µm
Supplier: Avantor
Silica gel 40 - 63 µm for flash chromatography
Supplier: VWR Chemicals
Reducing tubing connectors, b.safe
Supplier: Bohlender
VWR®, Crimp Caps for Crimp Neck Vials ND11
Supplier: VWR Collection
Empty disposable sample load cartridges, RediSep®
Supplier: COLLECTION PREFIX HPLC
HPLC columns, NUCLEODUR® NH2 / NH2-RP
Supplier: MACHEREY-NAGEL
Avantor® Alltima, HPLC Columns, C18
Supplier: Avantor
Capillaries for infinityLab quick connect fittings
Supplier: Agilent
Accessories for SCAT Safety Caps
Supplier: S.C.A.T.
Accessories for canisters, S60/61, with level indicator, b.safe
Supplier: Bohlender
20Mm Borate Buffer Ph9.3 100Ml 1 * 100 mL
Supplier: Agilent
Screw neck vials, N 15
Supplier: MACHEREY-NAGEL
Crimp caps, 20 mm, level 2 high-throughput applications, SureSTART™
Supplier: Thermo Fisher Scientific
Sample pans for autosamplers, Diamond™ DSC
Supplier: PerkinElmer
LC vials and closures
Supplier: Agilent