Order Entry
Ireland
ContactUsLinkComponent
LCMS Certified Amber Glass 12 x 32 mm Screw Neck Max Recovery Vial, with Cap and Preslit PTFE/Silicone Septum, 2 mL Volume, 1 * 100 items
Catalog # WATE600000755CV
Supplier:  WATERS
LCMS Certified Amber Glass 12 x 32 mm Screw Neck Max Recovery Vial, with Cap and Preslit PTFE/Silicone Septum, 2 mL Volume, 1 * 100 items
Catalog # WATE600000755CV
Supplier:  WATERS
Frequently Bought Together