3,4-DIHYDRO-7-HYDROXY-1H-QUINOLIN-2-ONE 1 * 5 g
Catalog # APOSOR59478-5G
Supplier: Apollo Scientific
3,4-DIHYDRO-7-HYDROXY-1H-QUINOLIN-2-ONE 1 * 5 g
Catalog # APOSOR59478-5G
Supplier: Apollo Scientific
Frequently Bought Together
Diethyl ether ≥99.7% stabilised, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
n-Hexane, Uvasol® for spectroscopy, Supelco®
Supplier: Merck
1,2-Dichloroethane 99+%, pure
Supplier: Thermo Fisher Scientific
N,N-Dimethylacetamide 99+%, extra pure
Supplier: Thermo Fisher Scientific
Methanol ≥99.9%, HiPerSolv CHROMANORM® for preparative HPLC
Supplier: VWR Chemicals
Ethyl acetate ≥99.5% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Xylene (mixture of isomers), J.T.Baker®
Supplier: Avantor
N,N-Dimethylacetamide 99%, pure
Supplier: Thermo Fisher Scientific
Petroleum spirit, 80…110 °C
Supplier: Roth Carl
(±)-1,2-Propanediol ACS
Supplier: Thermo Fisher Scientific
Dimethyl sulphoxide (DMSO),?99,9 %, Ph. Eur., USP 1 * 500 mL
Supplier: Roth Carl