4,4,4-TRIFLUORO-1-P-TOLYLBUTANE-1,3-DION 1 * 25 g
APOSPC200502-25G
Frequently Bought Together
ACETONITRILE EMPLURA® 1 * 4 L
: Merck
Ethyl acetate ≥99.5% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
: Honeywell Chemicals
N,N-Dimethylformamide 99+%, extra pure
: Thermo Fisher Scientific
Dimethyl sulphoxide, dried ≥99.9%, Puriss. p.a. (≤0.02% H₂O), Riedel-de Haën™
: Honeywell Chemicals
Benzyl alcohol ≥99%
: Thermo Fisher Scientific
Petroleum spirit, 30…40 °C, AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
Cesium chloride 99.99% (trace metals basis), Optical Grade for spectroscopy
: Thermo Fisher Scientific
Dichloromethane, extra dry 99.9% stabilised, AcroSeal®
: Thermo Fisher Scientific
Methanol ≥99.9%, CHROMASOLV™ for HPLC, Riedel-de Haën™
: Honeywell Chemicals
tert-Butyl methyl ether ≥99.0%, AnalaR NORMAPUR® ACS
: VWR Chemicals