METHYL 3-BROMO-2-(BROMOMETHYL)PROPANOATE 1 * 5 g
Catalog # APOSOR19510-5G
Supplier: Apollo Scientific
METHYL 3-BROMO-2-(BROMOMETHYL)PROPANOATE 1 * 5 g
Catalog # APOSOR19510-5G
Supplier: Apollo Scientific
Frequently Bought Together
1,2,3,4-Tetrahydronaphthalene for synthesis, Sigma-Aldrich®
Supplier: Merck
Pentane anhydrous, 99 % 1 * 1 L
Supplier: Merck
Cyclohexanone, EMPLURA®, extra pure, Supelco®
Supplier: Merck
(±)-2-Methyltetrahydrofuran 99+% stabilised, pure
Supplier: Thermo Fisher Scientific
Tetraheptylammonium bromide, Supelco®
Supplier: Merck
Acetonitrile 99.9%, ECD tested for residue analysis
Supplier: Thermo Fisher Scientific
Water with 0,1% (v/v) acetic acid, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Xylene - Mixture of isomers technical grade 1 * 5 L
Supplier: PanReac AppliChem
2-Propanol ≥99.5%, Laboratory Reagent
Supplier: Honeywell Chemicals
tert-Butyl methyl ether ≥99.0%, AnalaR NORMAPUR® ACS
Supplier: VWR Chemicals
Cyclohexanone ≥99.0% analytical reagent
Supplier: VWR Chemicals
Dimethyl sulphoxide, anhydrous ≥99.8%
Supplier: Thermo Fisher Scientific
Benzonitrile 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
Hydrochloric acid ≥36,5% ≥99.999% (metals basis)
Supplier: Thermo Fisher Scientific
Acetonitrile ≥99.9%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
n-Hexane 95%, ChromAR®, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
2-Propanol ≥99.8%, BAKER ANALYZED® for LC-MS, J.T.Baker®
Supplier: Avantor