2,2-DIFLUORO-2-PHENYLETHANAMINE HYDROCHL 1 * 250 mg
Catalog # APOSPC902092-250MG
Supplier: Apollo Scientific
2,2-DIFLUORO-2-PHENYLETHANAMINE HYDROCHL 1 * 250 mg
Catalog # APOSPC902092-250MG
Supplier: Apollo Scientific
Frequently Bought Together
Water with 0,1% (v/v) formic acid, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Petroleum spirit, 40…60 °C, PESTINORM® for capillary GC analysis
Supplier: VWR Chemicals
Acetonitrile, SupraSolv® for gas chromatography, Supelco®
Supplier: Merck
Dichloromethane stabilised, EMPLURA®
Supplier: Merck
Xylene (mixture of isomers) ≥98%, liquid
Supplier: PanReac AppliChem
Chloroform 99+% stabilised, extra pure
Supplier: Thermo Fisher Scientific
Tetrahydrofuran ≥99% stabilised
Supplier: Thermo Fisher Scientific
N,N-Dimethylformamide ≥99.9%, CHROMASOLV™ for headspace gas chromatography, Riedel-de Haën™
Supplier: Honeywell Chemicals
Isoamyl alcohol 98%, pure
Supplier: Thermo Fisher Scientific
2-Propanol ≥99.5%, Laboratory Reagent
Supplier: Honeywell Chemicals
(±)-2-Butanol, anhydrous 99%
Supplier: Thermo Fisher Scientific
ETHYLENE GLYCOL ≥98 TECHNICAL 1 * 5 L
Supplier: Roth Carl
n-Hexane ≥97%, HiPerSolv CHROMANORM® for preparative HPLC
Supplier: VWR Chemicals
Petroleum spirit, 40…60 °C for residue analysis, suitable for PAH analysis
Supplier: Thermo Fisher Scientific
2-Propanol ≥99.0%, GPR RECTAPUR®
Supplier: VWR Chemicals
2,2,4-Trimethylpentane, PRF B&J Brand, meets requirements of ASTM D2699 and D2700, Burdick & Jackson™
Supplier: Honeywell Chemicals