2-ETHYL-4-FLUOROPHENOL 1 * 5 g
APOSPC3232-5G
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Ethyl acetate ≥99%, TECHNICAL
: VWR Chemicals
Water USP, JP, Ph. Eur., purified
: VWR Chemicals
2-Propanol ≥99.0%, GPR RECTAPUR®
: VWR Chemicals
n-Heptane ≥99.0%, AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
: VWR Chemicals
Ethanol 70% ≥70% (v/v), TechniSolv®
: VWR Chemicals