Focusing lens for flexible light guides and gooseneck light guides up to 5 mm in diameter. 1 * 1 items
Catalog # LEIM10446391
Supplier: LEICA MICROSYSTEMS
Focusing lens for flexible light guides and gooseneck light guides up to 5 mm in diameter. 1 * 1 items
Catalog # LEIM10446391
Supplier: LEICA MICROSYSTEMS
Frequently Bought Together
VWR®, Microscope Covers
Supplier: VWR Collection
VWR® HCO 304, Conductivity Meter, Handheld
Supplier: VWR Collection
Accessories for Mettler balances
Supplier: Mettler - Toledo
VWR®, Accessories for Gravimetric Dilutors
Supplier: VWR Collection
VWR®, Accessories for SAS Super DUO 360
Supplier: VWR Collection
VWR® PH 20 pH Meter, Handheld
Supplier: VWR Collection
Accessories for cell counter, EVE™
Supplier: NANOENTEK
Accessories for Spectroquant® NOVA
Supplier: Merck
pH/mV/°C meters, handheld, pH 3110 / 3310
Supplier: WTW
VWR® pHenomenal® pH 1100 H pH/mV/°C Meter, Handheld
Supplier: VWR Collection
Short-term tubes for accuro®
Supplier: Dräger
OIL IMPACTOR 0.1-1.0 MG/M3 1 * 10 items
Supplier: Dräger
Plastic cuvette for LICO
Supplier: Hach
Accessories for buchi melting point apparatus M560 and M565
Supplier: BUCHI
Tubes and systems for active sampling
Supplier: Dräger
Gas detection tubes for sulphur dioxide
Supplier: Dräger
Accessories for titrators
Supplier: Mettler - Toledo