CALCITRIOL-D6 1 * 2,5 mg
Catalog # PROOTRCC144502-2.5
Supplier: LGC Standards PROMOCHEM
Frequently Bought Together
pH buffer solutions
Supplier: Mettler - Toledo
Calcium 1.000 mg/l in water standard for ion chromatography
Supplier: VWR Chemicals
VWR®, Chlorate 1,000 mg/L in Water standard for ion chromatography
Supplier: VWR Chemicals
pH buffers, DuraCal
Supplier: HAMILTON BONADUZ
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
pH buffers, ready to use
Supplier: WTW
VWR®, Sucrose (Brix) standards, stabilised solutions for refractometry
Supplier: VWR Chemicals
VWR®, Turbidity standards, formazin
Supplier: VWR Chemicals
Humidity standards
Supplier: ROTRONIC INSTRUMENTS
Benzophenone, Supelco®
Supplier: Merck
pH Standard Solutions, IUPAC, Certified
Supplier: Hach
Buffer solutions in sachets
Supplier: Mettler - Toledo