Carrier-free (BSA/glycerol-free) mouse monoclonal HSD17B13 Antibody ,Clone OTI5C4 1 * 100 G
ORIGCF814158
: OriGene
Carrier-free (BSA/glycerol-free) mouse monoclonal HSD17B13 Antibody ,Clone OTI5C4 1 * 100 G
ORIGCF814158
: OriGene
Frequently Bought Together
Eppendorf Tubes® 3810X Microtubes 1,5 ml
: EPPENDORF
Cryoboxes, 136×136 mm
: TENAK
Cryoboxes, 133×133 mm
: RATIOLAB
VWR® Standard Line Bottles, Narrow Neck, Round, PET, with Screw Caps
: VWR Collection
VWR®, Straight Sample Containers, with Screw Cap
: VWR Collection
Scoops
: VITLAB
Cell culture flasks, Corning®
: Corning
VWR®, Cryoboxes, Cardboard, 133×133 mm
: VWR Collection
Eppendorf twin.tec® PCR Plates 384-Well, 45 µl
: EPPENDORF
CryoStorage vials
: EPPENDORF
Seals for ALPS™ 50 V heat sealer
: THERMO MATRIX TECHNOLOGIES
Deep well plates, 96-well, SP Bel-Art
: Bel-Art Products, a Part of SP
Inoculating loops, stainless steel 18/10
: HAMMACHER KARL
Sample containers, with screw cap
: DELTALAB