Objective HC PL FL 63x/0.90 CORR Free working distance: 0.22 mm Correction collar for cover glass thickness of 0.11 - 0.23 mm 1 * 1 items
Catalog # LEIM11506223
Supplier: LEICA MICROSYSTEMS
Objective HC PL FL 63x/0.90 CORR Free working distance: 0.22 mm Correction collar for cover glass thickness of 0.11 - 0.23 mm 1 * 1 items
Catalog # LEIM11506223
Supplier: LEICA MICROSYSTEMS
Frequently Bought Together
Accessories for Pen-Ray® lamps
Supplier: UVP ULTRA VIOLET PRODUCTS
LED cold light sources, KL 1600 LED and KL 2500 LED
Supplier: SCHOTT AG Lighting and imaging
VWR®, UV Transilluminators for VWR® Imager System
Supplier: VWR Collection
Short-term tubes for accuro®
Supplier: Dräger
Flow Cells for MinION™ and GridION™ Devices
Supplier: Oxford Nanopore Technologies
Accessories for meters 765 / 766
Supplier: KNICK
VWR®, High Contrast Achromatic Objectives for VisiScope
Supplier: VWR Collection
Barbed luer adapters, Upchurch Scientific®
Supplier: UPCHURCH SCIENTIFIC
REINFORCED CLIP SHELF SS 250I 1 * 1 items
Supplier: Thermo Scientific
Accessories for VWR® V-1200, UV-1600PC, UV-3100PC
Supplier: VWR Collection
Evolution™ UV/VIS Spectrophotometers
Supplier: Thermo Fisher Scientific
Accessories for empty columns XK series
Supplier: Cytiva
Micro-Ostwald viscometers
Supplier: SI Analytics
AAA POWER CORD KIT EU 1 * 1 items
Supplier: Biochrom
Double-walled beaker 600 ml 1 * 1 items
Supplier: IKA
CONNECTOR LUER-TUBING 1,6MM ROTILABO 1 * 10 items
Supplier: Roth Carl