OBJECTIVE APOCHOMAT 0.32X S8APO 1 * 1 items
Catalog # LEIM10446334
Supplier: LEICA MICROSYSTEMS
Frequently Bought Together
VWR®, Accessories for VWR® INCU-Line® ILCO 180 PREMIUM
Supplier: VWR Collection
pH/mV/°C meters, handheld, Portavo® 902 and 904/904 ATEX
Supplier: KNICK
Hygiene monitoring system, HY-LiTE® 2
Supplier: Merck
Gas detection tube for hydrogen sulphide
Supplier: Dräger
VWR®, Heating Blocks, Aluminium, Designed for 1,5 ml Tubes
Supplier: VWR Collection
VWR® SAS Super DUO 360 Air Samplers
Supplier: VWR Collection
Pocket conductivity meters, LAQUAtwin, EC-11 / EC-22 / EC-33
Supplier: HORIBA
Conductivity meters, handheld, Portavo® 902 and 904/904 ATEX
Supplier: KNICK
Short-term tubes for accuro®
Supplier: Dräger
Accessories for Mettler balances
Supplier: Mettler - Toledo
Ubbelohde viscometers, uncalibrated
Supplier: SI Analytics
VWR® XT⁹⁶ XTender⁹⁶, Thermal Cyclers
Supplier: VWR Collection
Objectives for microscopes, Panthera TEC POL series
Supplier: MOTIC
Linen testers
Supplier: ESCHENBACH OPTIK
Luer-to-MicroTight® adapter, Upchurch Scientific®
Supplier: UPCHURCH SCIENTIFIC
Components for spectrophotometric colorimeters, PFX<i>i</i>-195/880/950/995 series, Lovibond®
Supplier: Lovibond Tintometer