TOTAL HARDNESS 1000 MG/L CALIBRATION ST 1 * 500 mL
ACSDHRDN1000.L5
: CPACHEM
Frequently Bought Together
Diethyl ether EMPROVE ESSENTIAL Ph Eur,BP 1 * 190 L
: MERCK PRODUCTION CHEMICALS
Glycerol 87% (BP, Ph. Eur.) pure, pharma grade 1 * 1.000 mL
: PanReac AppliChem
Diiodomethane 99+% stabilised
: Thermo Fisher Scientific
Chloroform ≥99.8% stabilised ACS, Riedel-de Haën™
: Honeywell Chemicals
Chloroform 99+% stabilised, extra pure
: Thermo Fisher Scientific
Sodium hexanitritocobaltate(III)
: Thermo Fisher Scientific
Acetonitrile ≥99%, TECHNICAL
: VWR Chemicals
1,5-Diphenylcarbazide, Supelco®
: Merck
Tetrahydrofuran ≥99.0% contains inhibitor, Reagent Grade
: Honeywell Chemicals
1,3-Dioxolane ≥99.0% stabilised, GPR RECTAPUR® for synthesis
: VWR Chemicals
1-Propanol ACS
: Thermo Fisher Scientific