C-KIT (V560G) ACTIVE 1 * 10 µG
Catalog # DISX14-730
Supplier: DISCOVERX
Frequently Bought Together
VWR®, Microcentrifuge Tubes and Screw Caps
Supplier: VWR Collection
VWR®, Petri Dish, PS
Supplier: VWR Collection
VWR®, Polyester Films for ELISA and General Incubation
Supplier: VWR Collection
Sampling spoons, sterile, Sterileware®, SP Bel-Art
Supplier: Bel-Art Products, a Part of SP
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
VWR®, Sample Dippers
Supplier: VWR Collection
VWR®, Microcentrifuge Tubes with Socket Screw Caps
Supplier: VWR Collection
Safe-Lock Tubes, 1,5 ml - Microtubes
Supplier: EPPENDORF
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR®, Cryoboxes, Cardboard, 133×133 mm
Supplier: VWR Collection
Spoon spatulas, LaboPlast® / SteriPlast®
Supplier: BURKLE
VWR®, Spreaders, L-shaped, Disposable
Supplier: VWR Collection
Screw caps for laboratory/media bottles
Supplier: SciLabware
PETRI DISH Ø 90 MM H 16MM VENT STERILE 1 * 500 items
Supplier: Anicrin
VWR®, Inoculating Loops and Needles, Disposable
Supplier: VWR Collection
VWR® Cryoboxes, Cardboard, 136×136 mm, with Full Labelling
Supplier: VWR Collection