PrecisION hGABAA 3/3/2 Recombinant Stable Cell Line 1 * 2 Vial
DISXCYL3068
Frequently Bought Together
Blender bags, BagLight®
: INTERSCIENCE
Eppendorf twin.tec® PCR Plates 96-well, 250 µl
: EPPENDORF
VWR®, Ice Pans, with or without Lid
: VWR Collection
Centrifuge tubes, Oak Ridge, Nalgene®
: Thermo Fisher Scientific
Cell culture flasks, Falcon®
: Corning
VWR®, Centrifuge Tubes, Conical Bottom, with Flat Screw Cap
: VWR Collection
Storage plates, 96-well, MASTERBLOCK®
: Greiner Bio-One
VWR®, Spreaders, L-Shaped, Standard Line
: VWR Collection
Cryoboxes, 133×133 mm
: TENAK
Cryoboxes, 136×136 mm
: RATIOLAB
Gel Load Pipette Tips
: Greiner Bio-One
Eppendorf® Assay/Reader Microplates
: EPPENDORF
VWR® Cryoboxes, Cardboard, 136×136 mm, with Full Labelling
: VWR Collection
Caps for roller bottles
: Corning