PathHunter CHO-K1 PTGER3 -Arrestin Cell Line 1 * 2 Vial
Catalog # DISX93-0640C2
Supplier: DISCOVERX
Frequently Bought Together
Sterilin™ Quickstart Universal Containers, PS, 30 ml
Supplier: STERILIN
Bioreactors, Wheaton® CELLine™
Supplier: DWK Life Sciences
Pasteur pipettes
Supplier: KARTELL
Microplate Sealing Mats
Supplier: EPPENDORF
Cryogenic tubes, CryoTubes™, Nunc™
Supplier: Thermo Fisher Scientific
Cryoboxes, B69/B50/B80/B99-Series
Supplier: GLW
VWR®, Cryoboxes, Cardboard, 133×133 mm
Supplier: VWR Collection
Serological pipettes, Stripette™
Supplier: Corning
Falcon® Petri Dishes, 50×9 mm, Tight-Fit Lid, Sterile, Corning
Supplier: Corning
Centrifuge tubes, 15 ml, Corning®
Supplier: Corning
Culture vessels with ports, with or without BioTech mixer, Nalgene®
Supplier: Thermo Fisher Scientific
Sterilin® Microtitre Plates, 96 Well
Supplier: STERILIN
Test tubes
Supplier: RATIOLAB
OmniTop Sample Tubes® Exact Volume Sampling System (EVSS)
Supplier: Avantor Fluid Handling
Collagen I-coated microplates, Corning® BioCoat®
Supplier: Corning
Storage plate, 48-well, 6,0 ml
Supplier: THERMO MATRIX TECHNOLOGIES
VWR® Cryoboxes, Cardboard, 136×136 mm, with Full Labelling
Supplier: VWR Collection
1536-Well cyclic olefin copolymer microplates, Corning®
Supplier: Corning