PathHunter CHO-K1 OXGR1 -Arrestin Orphan GPCR Cell Line 1 * 2 Vial
DISX93-0693C2
Frequently Bought Together
Transfer pipettes
: RATIOLAB
MicroWell™ Plates, 96 well, Nunc-Immuno™
: Thermo Fisher Scientific
VWR® Standard Line Transfer Pipettes
: VWR Collection
VWR® Blender Bags
: VWR Collection
Petri dishes
: Greiner Bio-One
VWR®, Transfer Pipettes, High Performance
: VWR Collection
Safe-Lock Tubes, 1,5 ml - Microtubes
: EPPENDORF
VWR®, Cryoboxes, Cardboard, 133×133 mm
: VWR Collection
Eppendorf® DNA LoBind® Microcentrifuge Tubes
: EPPENDORF
VWR®, Microcentrifuge Tubes, High Performance
: VWR Collection
VWR®, Conical Sample Containers, with Snap-On Lids
: VWR Collection
VWR® Serological Pipette, Standard Line
: VWR Collection
VWR®, Petri Dish, PS
: VWR Collection
VWR®, Straight Sample Containers, with Screw Cap
: VWR Collection
Deep well storage plates, 96-well, 0,8 ml
: THERMO MATRIX TECHNOLOGIES