PathHunter CHO-K1 mCRTH2 -Arrestin Cell Line 1 * 2 Vial
DISX93-0584C2
Frequently Bought Together
VWR® Premium Line, Serological Pipettes
: VWR Collection
Safe-Lock Tubes, 2,0 ml - Microtubes
: EPPENDORF
Eppendorf Tubes® 3810X Microtubes 1,5 ml
: EPPENDORF
VWR®, Spreaders, L-Shaped, Standard Line
: VWR Collection
VWR®, Straight Sample Containers, with Screw Cap
: VWR Collection
VWR®, Water Sampling Bottles, Sterile
: VWR Collection
VWR®, Strips of PCR Tubes and Caps
: VWR Collection
VWR® Ultra-High Performance Centrifuge Tubes
: VWR Collection
VWR®, Spreaders, T-shaped
: VWR Collection
VWR®, Cryoboxes, Cardboard, 136x136 mm
: VWR Collection
Cell culture flasks, Nunc™
: Thermo Fisher Scientific
VWR®, Sampling Spoons
: VWR Collection
VWR®, Cryoboxes, Cardboard, 133×133 mm
: VWR Collection
VWR®, Spreaders, L-shaped, Disposable
: VWR Collection
VWR®, Petri Dish, PS
: VWR Collection