PathHunter CHO-K1 C5AR1 -Arrestin Cell Line 1 * 2 Vial
Catalog # DISX93-0557C2
Supplier: DISCOVERX
Frequently Bought Together
Serological pipettes
Supplier: Greiner Bio-One
VWR®, Cryoboxes, Cardboard, 133×133 mm
Supplier: VWR Collection
FalconRegistered Mark Round-Bottom Tubes, 14 mL 1 * 500 items
Supplier: STEMCELL Technologies
Stoppers, Pierce-It™
Supplier: Simport Scientific
Microcentrifuge tubes, 0,5/1,5/2,0 ml
Supplier: Thermo Fisher Scientific
Hybridisation bottles, WHEATON®
Supplier: DWK Life Sciences
Freezer boxes (Cryoboxes), True North®
Supplier: HEATHROW
Internal thread cryogenic vials
Supplier: Corning
Cryoboxes, 136×136 mm
Supplier: RATIOLAB
Cryoboxes B13/B15/B25
Supplier: GLW
PCR Thermo-Tube and cap strips, 0.2 ml, ABgene®
Supplier: Thermo Fisher Scientific
BOTTLES HYBRIDIZATION 38 X 273 MM GL45 1 * 1 items
Supplier: witeg Labortechnik
Spare Lid of Ice Pan 1L, colour option, RED, BLUE, GREEN, BLACK 1 * 1 items
Supplier: VWR Collection