Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
AMINO ACIDS MIX STANDARD TYPE B (CRM) 1 * 5 mL
Catalog # WAKO011-27871
Supplier:
Wako
Expand image
AMINO ACIDS MIX STANDARD TYPE B (CRM) 1 * 5 mL
Catalog # WAKO011-27871
Supplier:
Wako
Frequently Bought Together
pH buffers, ready to use
Supplier: WTW
PIROCTONE OLAMINE 1 * 500 mg
Supplier: Merck
Hydrocortisone Impurity G Pharmaceutical Secondary Standard Certified Reference Material 1 * 50 mg
Supplier: Merck
Propoxur Analytical standard, reference material, Fluka™
Supplier: Honeywell Chemicals
Certified colour reference standards
Supplier: Lovibond Tintometer
Organic reference standard, Trichloroethylene 5000 µg/ml in methanol, Supelco®
Supplier: Merck
Polydisperse particle standards, CRM, Supelco®
Supplier: Merck
Supelco® Organic Reference Standard, 2-Isobutyl-3-methoxypyrazine solution
Supplier: Merck