0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Ireland
ContactUsLinkComponent
POLYMYXIN B SOLUTION 5X20ML 1 * 20 mL
HIMEA027-5X20ML
:
HIMEDIA
POLYMYXIN B SOLUTION 5X20ML 1 * 20 mL
HIMEA027-5X20ML
:
HIMEDIA
Frequently Bought Together
YEAST NITROGEN BASE, FOR MICROBIOLOGY* 1 * 100 g
: Merck
VWR® Culture Media, Ready to Use, in Bottles
: VWR Chemicals
MAC CONKEY MUG AGAR 1 * 500 g
: LIOFILCHEM
Antimicrobial susceptibility disks
: THERMO FISHER DIAGNOSTICS
CultiControl freeze-dried microorganism strains derived from NCTC® cultures
: LIOFILCHEM
VWR®, Pre-weighed dehydrated culture media in sachets
: VWR Chemicals
Antibiotic Sensitivity Discs
: HIMEDIA
MIDDLEBROOK 7H11 AGAR 20 plates 90 mm 1 * 1 items
: LIOFILCHEM