Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Recombinant human coronavirus SARS-CoV-2 Spike Glycoprotein RBD (mutated L452R) (Active) 1 * 50 µG
Catalog # ABCAAB283526-50
Supplier:
Abcam
Expand image
Recombinant human coronavirus SARS-CoV-2 Spike Glycoprotein RBD (mutated L452R) (Active) 1 * 50 µG
Catalog # ABCAAB283526-50
Supplier:
Abcam
Frequently Bought Together
VWR® Premium Line, Serological Pipettes
Supplier: VWR Collection
Individually Wrapped Serological Pipettes, Falcon®
Supplier: Corning
VWR® Multiwell Cell Culture Plates
Supplier: VWR Collection
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Metal-Free, with Screw Cap
Supplier: VWR Collection
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
Eppendorf® Protein LoBind® Microcentrifuge Tubes
Supplier: EPPENDORF
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Safe-Lock Tubes, 1,5 ml - Microtubes
Supplier: EPPENDORF
VWR®, Cryoboxes, Cardboard, 133×133 mm
Supplier: VWR Collection
VWR®, Cryoboxes with Dividers
Supplier: VWR Collection