Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
BOOT JALAS 2128 VIP SAFETY EU38/UK5 1 * 1 PAIR
Catalog # EJEN2128-38
Supplier:
EJENDALS
Expand image
BOOT JALAS 2128 VIP SAFETY EU38/UK5 1 * 1 PAIR
Catalog # EJEN2128-38
Supplier:
EJENDALS
Frequently Bought Together
Nitrile gloves, TouchNTuff® 92-600 / 92-605
Supplier: Ansell
Neoprene gloves, Microflex® NeoTouch® 25-101 / 25-201
Supplier: Ansell
Colour Lab Tapes
Supplier: GA
Nitrile gloves, Kimtech™ Purple Nitrile™
Supplier: KIMBERLY CLARK
VWR®, Latex Gloves
Supplier: VWR Collection
Nitrile gloves, TouchNTuff® 92-665 / 92-670
Supplier: Ansell
VWR®, Nitrile Gloves
Supplier: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
Supplier: VWR Collection
Cleanroom Gloves, Kimtech Pure™ G3 NxT™
Supplier: KIMBERLY CLARK
VWR® Labels, Hazardous Substance, GHS Labels
Supplier: VWR Collection