ScienceLine plus pH combination electrode 3 m fixed cable with DIN and banana plug, ID-funct., temperature sensor Pt 1000, glass shaft, Pt junction, electrolyte KCl 3 mol/l, Silamid®-reference system with silver ion barrier and double-electrolyte, sp 1 * 1 items
Catalog # SCOI285101320
Supplier: SI Analytics
Frequently Bought Together