HIV-1 p24 (group M, subtype B, strain 92418) Antibody, Mouse MAb 1 * 100 µl
Catalog # SINO11695-MM15-100
Supplier: Sino Biological
HIV-1 p24 (group M, subtype B, strain 92418) Antibody, Mouse MAb 1 * 100 µl
Catalog # SINO11695-MM15-100
Supplier: Sino Biological
Frequently Bought Together
Sampling spoons with long handle, SP Bel-Art
Supplier: Bel-Art Products, a Part of SP
VWR® Standard Line Bottles, Narrow Neck, Round, PET, with Screw Caps
Supplier: VWR Collection
VWR®, Cryoboxes, Cardboard, 133×133 mm
Supplier: VWR Collection
Scoops
Supplier: LAMAPLAST
VWR®, Blotting Pads, Grade 707
Supplier: VWR Collection
Cell culture inserts for multiwell plates, ThinCert™
Supplier: Greiner Bio-One
VWR® Cell Chambers
Supplier: VWR Collection
CostarRegistered Mark 6.5 mm TranswellRegistered Mark, 0.4 µm Pore Polyester Membrane Inserts 1 * 48 items
Supplier: STEMCELL Technologies
PCR microtube and cap strips, 0.2 ml, 8-strips, Axygen
Supplier: Corning
Vials for environmental analysis, WHEATON®
Supplier: DWK Life Sciences
Cap sealing mats, Matrix SepraSeal® and DuraSeal®
Supplier: THERMO MATRIX TECHNOLOGIES
Racks for Upright Freezers with Drawers
Supplier: GLW
Accessories for conical-bottom bottles, NALGENE®
Supplier: Thermo Fisher Scientific
VWR® Cryoboxes, Cardboard, 136×136 mm, with Full Labelling
Supplier: VWR Collection