Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Recombinant Anti-Cystatin SN Antibody, Rabbit Monoclonal 1 * 100 µl
Catalog # SINO11568-R111-100
Supplier:
Sino Biological
Expand image
Recombinant Anti-Cystatin SN Antibody, Rabbit Monoclonal 1 * 100 µl
Catalog # SINO11568-R111-100
Supplier:
Sino Biological
Frequently Bought Together
Transfer pipettes
Supplier: RATIOLAB
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
Cell culture flasks, Falcon®
Supplier: Corning
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR®, Spreaders, L-shaped, Disposable
Supplier: VWR Collection
Safe-Lock Tubes, 1,5 ml - Microtubes
Supplier: EPPENDORF
Safe-Lock Tubes, 2,0 ml - Microtubes
Supplier: EPPENDORF
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
Centrifuge tubes, conical bottom, Falcon®
Supplier: Corning
VWR®, Centrifuge Tubes, Conical Bottom
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Freestanding, with Flat Screw Caps
Supplier: VWR Collection
VWR®, Cell Culture Flasks, Non Treated or Treated
Supplier: VWR Collection