Anti-DUSP3 Antibody, Rabbit Polyclonal 1 * 50 µl
SINO10114-T52-50
Frequently Bought Together
VWR®, Cryoboxes, Cardboard, 136x136 mm
: VWR Collection
Cell culture inserts, Falcon®
: Corning
Scoops for foodstuffs, SteriPlast®
: BURKLE
DURAN® Reactor, Stirred, GLS 80 Thread
: DWK Life Sciences
Ice buckets, Bel-Art Magic Touch 2™
: Bel-Art Products, a Part of SP
Roller bottle standard, surface treated, 1000 ml vent cap 1 * 24 items
: GUANGZHOU JET BIOFILTRATION
Cryoboxes, 133×133 mm
: RATIOLAB
Tubes Cryo.S™ 4 and 5 ml
: Greiner Bio-One
Sample containers, straight, with screw cap, WHEATON®
: DWK Life Sciences
Microplates, 96-well, Nunc™ Edge 2.0
: Thermo Fisher Scientific
Gel blotting sheets, GB005, Whatman™
: Whatman products (Cytiva)