Recombinant Anti-CD14 Antibody (PerCP), Rabbit Monoclonal 1 * 100 Tests
Catalog # SINO10073R002C-100
Supplier: Sino Biological
Recombinant Anti-CD14 Antibody (PerCP), Rabbit Monoclonal 1 * 100 Tests
Catalog # SINO10073R002C-100
Supplier: Sino Biological
Frequently Bought Together
Transfer pipettes
Supplier: RATIOLAB
VWR®, Spreaders, T-shaped
Supplier: VWR Collection
Eppendorf Tubes® 3810X Microtubes 1,5 ml
Supplier: EPPENDORF
VWR® Premium Line, Serological Pipettes
Supplier: VWR Collection
Centrifuge tubes, 15 ml, Corning®
Supplier: Corning
VWR® Water Sampling Bottles, with Tamper Evident Cap
Supplier: VWR Collection
VWR®, Petri Dish, PS
Supplier: VWR Collection
Eppendorf Tubes® Microtubes with Snap Cap, 5,0 ml
Supplier: EPPENDORF
Eppendorf® Protein LoBind® Microcentrifuge Tubes
Supplier: EPPENDORF
VWR®, Centrifuge Tubes, High Performance, Flat or Plug Cap, PP
Supplier: VWR Collection
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
Serological pipettes, Stripette™
Supplier: Corning
VWR®, Conical Sample Containers, with Snap-On Lids
Supplier: VWR Collection
VWR® Multiwell Cell Culture Plates
Supplier: VWR Collection
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
Supplier: VWR Collection
VWR®, Cryoboxes, Cardboard, 133×133 mm
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection