0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Ireland
ContactUsLinkComponent
Recombinant Cynomolgus IL-21 Receptor/IL-21R Protein (His Tag) 1 * 100 µG
SINO90315-C08H-100
:
Sino Biological
Recombinant Cynomolgus IL-21 Receptor/IL-21R Protein (His Tag) 1 * 100 µG
SINO90315-C08H-100
:
Sino Biological
Frequently Bought Together
Centrifuge tubes, conical bottom, Falcon®
: Corning
VWR® Inoculating Loops and Needles, Disposable
: VWR Collection
VWR® Serological Pipette, Standard Line
: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
: VWR Collection
VWR®, Straight Sample Containers, with Screw Cap
: VWR Collection
Transfer pipettes
: RATIOLAB
VWR® Premium Line, Serological Pipettes
: VWR Collection
Safe-Lock Tubes, 2,0 ml - Microtubes
: EPPENDORF
VWR® Standard Line Transfer Pipettes
: VWR Collection
Safe-Lock Tubes, 1,5 ml - Microtubes
: EPPENDORF
VWR®, Transfer Pipettes, High Performance
: VWR Collection
Petri dishes
: Greiner Bio-One
VWR®, Spreaders, L-shaped, Disposable
: VWR Collection