Recombinant Canine FOLR1 Protein (His Tag) 1 * 100 µG
Catalog # SINO70115-D08H-100
Supplier: Sino Biological
Recombinant Canine FOLR1 Protein (His Tag) 1 * 100 µG
Catalog # SINO70115-D08H-100
Supplier: Sino Biological
Frequently Bought Together
Amersham™ Protran® Western Blotting Membranes, Nitrocellulose
Supplier: Cytiva
Cooling systems, CoolSafe™
Supplier: DIVERSIFIED BIOTECH
MICRO TUBE 2.0ML STERILE SKIRTED TYPE I 1 * 1.000 items
Supplier: SARSTEDT
Pasteur Pipette,Extended Fine Tip,3.3ml Sterile 1 * 500 items
Supplier: DELTALAB
96-well solid black and white polystyrene microplates, Corning®
Supplier: Corning
Cell culture roller bottles, Corning®
Supplier: Corning
Portable Vaccine Coolers
Supplier: GLEN DIMPLEX
Storage tubes, 2D barcoded, Matrix™
Supplier: THERMO MATRIX TECHNOLOGIES
PCR racks for tubes, strips and plates, PCRack™
Supplier: Simport Scientific
VESSEL REACT 0.5 ML PP PROTEIN LOW 1 * 1.200 items
Supplier: SARSTEDT