Recombinant Mouse TSLP Protein (His Tag) 1 * 100 µG
Catalog # SINO51005-M08H-100
Supplier: Sino Biological
Recombinant Mouse TSLP Protein (His Tag) 1 * 100 µG
Catalog # SINO51005-M08H-100
Supplier: Sino Biological
Frequently Bought Together
VWR® Square Polycarbonate Media Bottles
Supplier: VWR Collection
VWR®, Cryoboxes, Cardboard, 136x136 mm
Supplier: VWR Collection
Transfer pipettes, graduated, Samco™
Supplier: SAMCO
Storage Boxes, PP, with Lid
Supplier: KOEHLER TECHNISCHE PRODUKTEN
Caps for microcentrifuge tubes with thread
Supplier: Simport Scientific
Carboys, narrow neck, PC, square, Biotainer™, Nalgene®
Supplier: Thermo Fisher Scientific
Transfer pipettes, fine tip
Supplier: Alpha Laboratories
SENSOPLATE 24W GLASS BOTTOM W/O LID ST 1 * 12 items
Supplier: Greiner Bio-One
Pierce™ Immunoassay Microplates, Steptavidin Coated, 96-Well
Supplier: Thermo Fisher Scientific
Storage tubes, 2D barcoded, Matrix™
Supplier: THERMO MATRIX TECHNOLOGIES
Cell culture flasks, Nunc™ TripleFlask™
Supplier: Thermo Fisher Scientific