SIGIRR Protein, Human, Recombinant (His Tag) 1 * 1 mg
Catalog # SINO12165-H08H-1
Supplier: Sino Biological
SIGIRR Protein, Human, Recombinant (His Tag) 1 * 1 mg
Catalog # SINO12165-H08H-1
Supplier: Sino Biological
Frequently Bought Together
Sterilin™ Quickstart Universal Containers, PS, 30 ml
Supplier: STERILIN
Transfer pipettes
Supplier: RATIOLAB
VWR®, Centrifuge Tubes, High Performance, Flat or Plug Cap, PP
Supplier: VWR Collection
Petri dishes, square, Corning® Gosselin™
Supplier: Corning
Cap strips, flat top
Supplier: Thermo Fisher Scientific
Cryogenic tubes, CryoTubes™, Nunc™
Supplier: Thermo Fisher Scientific
Serological pipettes, Stripette™
Supplier: Corning
Inoculating loops with needle end
Supplier: Brand
VWR®, Sample Containers, with Attached Lids
Supplier: VWR Collection
VWR® Premium Line, Serological Pipettes
Supplier: VWR Collection
Safe-Lock Tubes, 1,5 ml - Microtubes
Supplier: EPPENDORF
VWR®, Cryoboxes, Cardboard, 133×133 mm
Supplier: VWR Collection
Round-bottom polystyrene tubes, disposable, Falcon®
Supplier: Corning
VWR®, Centrifuge Tubes, Freestanding, with Flat Screw Caps
Supplier: VWR Collection
Sealing tape for MicroWell™ plates, Nunc™
Supplier: Thermo Fisher Scientific
VWR®, Straight Sample Containers, with Screw Cap
Supplier: VWR Collection
Eppendorf Tubes® Microtubes with Snap Cap, 5,0 ml
Supplier: EPPENDORF
Eppendorf® Protein LoBind® Microcentrifuge Tubes
Supplier: EPPENDORF