Anti-Pentraxin 3 Antibody, Rabbit Polyclonal 1 * 400 µl
Catalog # SINO12082-RP01-400
Supplier: Sino Biological
Anti-Pentraxin 3 Antibody, Rabbit Polyclonal 1 * 400 µl
Catalog # SINO12082-RP01-400
Supplier: Sino Biological
Frequently Bought Together
Blender bags, BagLight®
Supplier: INTERSCIENCE
VWR® Cryoboxes, Cardboard, 136×136 mm, with Full Labelling
Supplier: VWR Collection
VWR®, Cryoboxes, Cardboard, 133×133 mm
Supplier: VWR Collection
PCR tubes, 0.2 ml, with flat or domed cap, ABgene®
Supplier: Thermo Fisher Scientific
Containers with snap caps, Corning® Gosselin™
Supplier: Corning
MULTICELL HOLDER 8-POSITION 1 * 1 items
Supplier: PerkinElmer
Cryogenic Vials with Internal Thread, 1.8 mL 1 * 500 items
Supplier: STEMCELL Technologies
PCR tube strips with attached caps, EasyStrip™ Plus
Supplier: Thermo Fisher Scientific
OmniTop Sample Tubes® Exact Volume Sampling System (EVSS)
Supplier: Avantor Fluid Handling
VWR® Safe-T-Flex, Caps for Test Tubes
Supplier: VWR Collection
Containers, straight, wide mouth, with screw cap, Corning® Gosselin™
Supplier: Corning