Recombinant Human SLAMF6 Protein (His Tag) 1 * 1 mg
Catalog # SINO11945-H08H-1
Supplier: Sino Biological
Recombinant Human SLAMF6 Protein (His Tag) 1 * 1 mg
Catalog # SINO11945-H08H-1
Supplier: Sino Biological
Frequently Bought Together
Safe-Lock Tubes, 2,0 ml - Microtubes
Supplier: EPPENDORF
Cryoboxes, 133×133 mm
Supplier: TENAK
Cryogenic tubes
Supplier: Brand
Microplate stands
Supplier: DIVERSIFIED BIOTECH
Corning® 1536 well cyclic olefin copolymer (COC) microplate
Supplier: Corning
Screw caps for micro tubes
Supplier: SARSTEDT
Cell culture inserts, Falcon®
Supplier: Corning
Storage Boxes
Supplier: EPPENDORF
Microplates, 96- and 384-well, UV Star®
Supplier: Greiner Bio-One
Measuring scoops
Supplier: VITLAB
Freezer boxes (Cryoboxes), True North®
Supplier: HEATHROW
PIPETTE PASTEUR ULTRA MICRO 25 DROPS/ML 1 * 500 items
Supplier: Alpha Laboratories
Containers, straight, wide mouth, with screw cap, Corning® Gosselin™
Supplier: Corning
Poly-D-lysine plates, Corning® BioCoat®
Supplier: Corning
Cryostorage boxes, KeepIT-100™, WHEATON®
Supplier: DWK Life Sciences