Recombinant Human interferon-alpha/IFNA5 Protein (His Tag) 1 * 100 µG
SINO10342-H08Y-100
Recombinant Human interferon-alpha/IFNA5 Protein (His Tag) 1 * 100 µG
SINO10342-H08Y-100
Frequently Bought Together
Safe-Lock Tubes, 2,0 ml - Microtubes
: EPPENDORF
Eppendorf Tubes® 3810X Microtubes 1,5 ml
: EPPENDORF
VWR® Blender Bags
: VWR Collection
VWR® Standard Line Transfer Pipettes
: VWR Collection
Cell culture flasks, Falcon®
: Corning
Transfer pipettes
: RATIOLAB
Safe-Lock Tubes, 1,5 ml - Microtubes
: EPPENDORF
VWR®, Transfer Pipettes, High Performance
: VWR Collection
VWR®, Spreaders, L-shaped, Disposable
: VWR Collection
Serological pipettes, Stripette™
: Corning
VWR® Serological Pipette, Standard Line
: VWR Collection
VWR®, Straight Sample Containers, with Screw Cap
: VWR Collection
VWR®, Petri Dish, PS
: VWR Collection
VWR® Standard Line Bottles, Narrow Neck, Round, PET, with Screw Caps
: VWR Collection